Corning® SSPE, Liquid, 20X, Corning
Catalog # 45001-128
Supplier: Corning
CAS Number:
Specifications
- Description:20X SSPE
- Size:6×1 L
- Storage Temperature:Ambient
- Shipping Temperature:Ambient
- Cat. No.:45001-128
- Form:Liquid
- Supplier no.:46-021-CM
Specifications
About this item
20X Saline-Sodium-Phosphate-EDTA (SSPE) buffer has applications in nucleic acid hybridization and other molecular biology work.