Order Entry
Puerto Rico
ContactUsLinkComponent
Corning® Tris, 10X Buffered Saline, Liquid, Corning
Corning® Tris, 10X Buffered Saline, Liquid, Corning
Catalog # 45001-132
Supplier:  Corning
CAS Number:  
undefined
Corning® Tris, 10X Buffered Saline, Liquid, Corning
Catalog # 45001-132
Supplier:  Corning
Supplier Number:  46-012-CM
CAS Number:  

Specifications

  • Pack type:
    6/pack
  • Description:
    Tris, Corning® 1 L 10X Tris buffered saline, liquid, pH 7.4 ±0.1 RNase-/DNase- and protease-free
  • Size:
    1 L
  • Storage Temperature:
    Ambient
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    45001-132
  • Supplier no.:
    46-012-CM

Specifications

About this item

Tris Buffered Saline (TBS) is an isotonic, non-toxic buffer used in some biomedical techniques to maintain pH within a relatively narrow range

TBS can shift pH relatively dramatically as temperature changes, with the pKa declining approximately 0.03 units per degree Celsius rise in temperature.