Order Entry
Puerto Rico
ContactUsLinkComponent
Corning® 10% SDS (Sodium Dodecyl Sulfate), Corning
Corning® 10% SDS (Sodium Dodecyl Sulfate), Corning
Catalog # 45001-120
Supplier:  Corning
CAS Number:  
undefined
Corning® 10% SDS (Sodium Dodecyl Sulfate), Corning
Catalog # 45001-120
Supplier:  Corning
Supplier Number:  46-040-CI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    10% SDS (Sodium Dodecyl Sulfate)
  • Size:
    100 mL
  • Storage Temperature:
    Ambient
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    45001-120
  • Supplier no.:
    46-040-CI

Specifications

About this item

Sodium Dodecyl Sulfate (SDS) is an anionic detergent with applications in SDS-PAGE protein analysis and nucleic acid purification. It is a wetting agent that is effective in both acid and alkaline solutions, and is most often used as a protein and lipid solubilization reagent. SDS is also a powerful protein denaturant. This 10% solution is prepared in 18 megaohm water and is 0.2µm filtered.