Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
β-Cyclodextrine, Reagent Grade suitable for use as excipient

β-Cyclodextrine, Reagent Grade suitable for use as excipient

β-Cyclodextrine, Reagent Grade suitable for use as excipient
Certificates
β-Cyclodextrine, Reagent Grade suitable for use as excipient

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

beta-Cyclodextrin, Reagent is cyclic polysaccharide comprised of 7 glucopyranose units. This ingredient appears as a solid and functions as a chelating agent. Cyclodextrins are able to form host-guest complexes with hydrophobic molecules and are used in the production cholesterol free products. Beta-cyclodextrin complexes with certain carotenoid food colorants have been shown to intensify color, increase water solubility and improve light stability. Polymeric microspheres made with beta-cyclodextrin show high oil-absorbancy. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Synonyms:  Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
  • CAS Number:7585-39-9
  • Formula:C₄₂H₇₀O₃₅
  • MDL Number:MFCD00078139
  • Molecular Weight:1134.98 g/mol
  • Storage Temperature:Ambient
  • EINECS:231-493-2

Documents

Certificates
Search by Lot Number
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass, amber
    5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $270.36
    $270.36
    Each (5g)
       $270.36

    Supplier Number: C1965-5GM
  • Glass, amber
    25 g
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $502.92
    $502.92
    Each (25g)
       $502.92

    Supplier Number: C1965-25GM

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.