Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
γ-Cyclodextrine ≥98% suitable for use as excipient

γ-Cyclodextrine ≥98% suitable for use as excipient

γ-Cyclodextrine ≥98% suitable for use as excipient
Certificates
γ-Cyclodextrine ≥98% suitable for use as excipient

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

gamma-Cyclodextrin, also known as Cyclodextrins, are used in the food, pharmaceutical, and chemical industries because they enhance the solubility and bioavailability of such compounds. This is of high interest for pharmaceutical as well as dietary supplement applications in which hydrophobic compounds shall be delivered. Alpha-, beta-, and gamma-cyclodextrin are all generally recognized as safe by the FDA.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Synonyms:  gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
  • CAS Number:17465-86-0
  • Formula:C₄₈H₈₀O₄₀
  • MDL Number:MFCD00009595
  • Molecular Weight:1297.1248 g/mol
  • Storage Temperature:Ambient
  • EINECS:241-482-4

Documents

Certificates
Search by Lot Number
Specifications for Product Family

Specification Test Results

  • Assay (%):
    98

Specifications

Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass, amber
    100 mg
    Log in for availability
    Log in for availability
    Each (100mg)
    Each (100mg) - $171.88
    $171.88
    Each (100mg)
       $171.88

    Supplier Number: C1623-100MG
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    700011-306
    Small Business Enterprise
  • Glass, amber
    1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $506.26
    $506.26
    Each (1g)
       $506.26

    Supplier Number: C1623-1GM
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    700011-308
    Small Business Enterprise
  • Glass, amber
    5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $1,934.34
    $1,934.34
    Each (5g)
       $1,934.34

    Supplier Number: C1623-5GM
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    700011-310
    Small Business Enterprise

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.