Order Entry
Puerto Rico
ContactUsLinkComponent
N-[2-(N-Vinylbenzylamino)ethyl]-3-aminopropyltrimethoxysilane hydrochloride 30 - 40% in methanol
N-[2-(N-Vinylbenzylamino)ethyl]-3-aminopropyltrimethoxysilane hydrochloride 30 - 40% in methanol
Catalog # TCV0050-025ML
Supplier:  TCI America
CAS Number:  
N-[2-(N-Vinylbenzylamino)ethyl]-3-aminopropyltrimethoxysilane hydrochloride 30 - 40% in methanol
Catalog # TCV0050-025ML
Supplier:  TCI America
Supplier Number:  V0050-25ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    25 mL
  • Cat. No.:
    TCV0050-025ML
  • Supplier no.:
    V0050-25ML

Specifications

About this item

CAS Number:34937-00-3
 :
Formula:C₁₇H₃₀N₂O₃Si
MDL Number:MFCD00060149
 :374.98 g/mol
Flash Pt:12 °C