Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Thioflavin T

Thioflavin T

25 g
Catalog # TCT0558-025G
Supplier:   TCI America
Supplier #:  T0558-25G
Thioflavin T
Catalog # TCT0558-025G
Supplier:  TCI America
CAS Number:  2390-54-7
Thioflavin T
Thioflavin T

25 g

CAS Number:  2390-54-7
Price
$47.00
Log in for availability
Each (25g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 2390-54-7 MDL Number: MFCD00011944 Molecular Formula: C17H19ClN2S Molecular Weight: 318.86 Form: Crystal Color: Yellow

Form: Crystal
Color: Yellow

Synonyms:  CI 49005, Thioflavine T, Basic Yellow 1, 2-(4-(Dimethylamino)phenyl)-3,6-dimethylbenzo[d]thiazol-3-ium chloride, Setoflavine T
  • CAS Number:2390-54-7
  • Formula:C₁₇H₁₉ClN₂S
  • MDL Number:MFCD00011944
  • Molecular Weight:318.86 g/mol

Documents

Specifications

Specifications

  • Size:
    25 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.