Order Entry
Puerto Rico
ContactUsLinkComponent
Reference Material of Hydrocarbon Types in Gasoline by FIA Certified by The Japan Petroleum Institute, TCI America
Reference Material of Hydrocarbon Types in Gasoline by FIA Certified by The Japan Petroleum Institute, TCI America
Catalog # TCS0806-SET
Supplier:  TCI America
CAS Number:  
Reference Material of Hydrocarbon Types in Gasoline by FIA Certified by The Japan Petroleum Institute, TCI America
Catalog # TCS0806-SET
Supplier:  TCI America
Supplier Number:  S0806-1SET
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    Set of 3×5 ml
  • Cat. No.:
    TCS0806-SET
  • Contents:
    Olefins Content 25vol% level
  • Supplier no.:
    S0806-SET

Specifications

About this item

Class 3