Order Entry
Puerto Rico
ContactUsLinkComponent
Methyl Nonanoate, Standard Material for GC
Methyl Nonanoate, Standard Material for GC
  TCS0307-005ML
 :  
Methyl Nonanoate, Standard Material for GC
  TCS0307-005ML
 :  S0307-5ML
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Pack type:
    Glass bottle
  • CAS No.:
    1731-84-6
  • Size:
    5 ml
  • Formula:
    C₁₀H₂₀O₂
  • Description:
    Methyl Nonanoate
  • Flash Point:
    87 °C (lit.)
  • Boiling Point:
    214 °C (lit.)
  • Appearance:
    Colorless clear liquid
  • Cat. No.:
    TCS0307-005ML
  • Contents:
    Methyl Nonanoate, Purity (GC): min. 99.5 %
  • Supplier no.:
    S0307-5ML

 

 

Specific Gravity (20/20): 0.88

Product Categories: Analytical Science, Fatty Acid Methyl Esters (GC Standard) Other Product Names / Synonyms: Methyl Pelargonate, Nonanoic Acid Methyl Ester, Pelargonic Acid Methyl Ester