Order Entry
Puerto Rico
ContactUsLinkComponent
 
Neu5GcAc[1Me,4789Ac]α(2-6)Gal[24Bz,3Bn]-β-MP ≥97.0% (by HPLC)
Catalog # TCM1763-200MG
Supplier:  TCI America
CAS Number:  
Neu5GcAc[1Me,4789Ac]α(2-6)Gal[24Bz,3Bn]-β-MP ≥97.0% (by HPLC)
Catalog # TCM1763-200MG
Supplier:  TCI America
Supplier Number:  M1763-200MG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    200 mg
  • Cat. No.:
    TCM1763-200MG
  • Supplier no.:
    M1763-200MG

Specifications

Specification Test Results

  • Purity/Analysis Method:
    >97.0% (HPLC)

Specification Test Results

About this item

 :
 :1116.09 g/mol
Formula:C₅₆H₆₁NO₂₃