Diethyl-trans-3-hexenedioate ≥98.0%
Product Details & Documents
CAS Number: 57042-08-7
MDL Number: MFCD00191608
Molecular Formula: C10H16O4
Molecular Weight: 200.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.04
Other Product Names / Synonyms: trans-2-Butene-1,4-dicarboxylic Acid Diethyl Ester, trans-3-Hexenedioic Acid Diethyl Ester, Diethyl trans-2-Butene-1,4-dicarboxylate
- CAS Number:57042-08-7
- Formula:C₁₀H₁₆O₄
- MDL Number:MFCD00191608
- Molecular Weight:200.23 g/mol
- Density:1.04 (20 °C)
Specifications
Specification Test Results
- Purity/Analysis Method:>98.0% (GC)
Specifications
- Size:10 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



