Order Entry
Puerto Rico
ContactUsLinkComponent
Lead(II) acetate 10% (w/v) in 2% acetic acid
Lead(II) acetate 10% (w/v) in 2% acetic acid
Catalog # RCSOD20-32
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Lead(II) acetate 10% (w/v) in 2% acetic acid
Catalog # RCSOD20-32
Supplier:  Ricca Chemical
Supplier Number:  RSOD2000-1A
Small Business Enterprise
CAS Number:  

Specifications

  • Size:
    32 oz.
  • Cat. No.:
    RCSOD20-32
  • Supplier no.:
    RSOD2000-1A

Specifications

About this item

 :
 :325.29 g/mol
UN:1616
ADR:6.1,III
CAS Number:301-04-2
Formula:(H₃CCOO)₂Pb
MDL Number:MFCD00012452