Order Entry
Puerto Rico
ContactUsLinkComponent
Capping Reagent (Acetic Anhydride:Lutidine:Acetonitrile) (20:30:50), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Irritant
Corrosive
Health
Flammable
Capping Reagent (Acetic Anhydride:Lutidine:Acetonitrile) (20:30:50), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Catalog # BJSR644-20
Supplier:  Burdick & Jackson
CAS Number:  
Irritant
Corrosive
Health
Flammable
Capping Reagent (Acetic Anhydride:Lutidine:Acetonitrile) (20:30:50), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Catalog # BJSR644-20
Supplier:  Burdick & Jackson
Supplier Number:  SR644-20
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Steel NOWPAK®
  • Size:
    20 L
  • Cat. No.:
    BJSR644-20
  • Supplier no.:
    SR644-20

Specifications

About this item

 :
Density:0.881 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
UN:2924
ADR:6.1,II
 :GHS07
 :GHS05
 :GHS08
 :GHS02