

2-Propanol:Water solution (50:50)% (v/v) for LC-MS, for flushing LC-MS systems, Burdick & Jackson™
Catalog # BJLC703-1
Supplier: Honeywell Research Chemicals
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Glass bottle, GL45
- Size:1 L
- Cat. No.:BJLC703-1
- Supplier no.:LC703-1
Specifications
About this item
:
Boiling Pt:82.3 °C (solvent)
Density:0.8 to 0.92 g/cm³ (20 °C)
Flash Pt:22.78 °C
Flash Pt:12
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
:GHS02
:GHS07
:7732-18-5