ContactUsLinkComponent
Transport Cart for 200 L Chemical Containers, Honeywell B&J
Transport Cart for 200 L Chemical Containers, Honeywell B&J
Catalog # BJ-CA2375
Product available on GSA Advantage®
Transport Cart for 200 L Chemical Containers, Honeywell B&J
Catalog # BJ-CA2375
Supplier Number:  CA2375
Product available on GSA Advantage®
Specifications
  • Description:
    Cart for 200 L SSPDS container
  • Cat. No.:
    BJ-CA2375
  • Supplier no.:
    CA2375
Specifications
About this item
The carts are specially designed to conveniently and safely move 200 L SSPDS containers around tight areas and over uneven surfaces such as door frames and ramps
includes hand break.