Drench Hose Attachment, Speakman®
Catalog # VF-4920-SE
Supplier: Speakman
Small Business Enterprise
Specifications
- Color:Yellow and White
- Description:Drench Hose Attachment
- Material:Stainless Steel, Brass, Plastic, and Steel Composition
- Cat. No.:VF-4920-SE
- Supplier no.:SE-4920
Specifications
About this item
The drench hose attachment is designed for use with the Speakman® portable GravityFlo® units.
This attachment allows for spot body washing, in addition to the unit's eyewash flushing capability.