Axygen® IMAG Hand-Held Magnetic Separation Device, Corning
Supplier: Corning
Certificates
About this item
The magnetic separation devices (MSD) are essential in any paramagnetic bead-based purification process. These IMAG™ devices are designed for a manual process.
- Single tube or 96-well format
- Applications include nucleic acid purification, cell based assays and antobody and protein purifications
- Strong magnets enable separation times of less than 30 seconds
- Manual oeration for either low or high throughput
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 97067-594
- 97067-596
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental Certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa