ContactUsLinkComponent
SP Bel-Art Gallon Bottle, Wide-Mouth, Bel-Art Products, a part of SP
  16126-764
undefined
SP Bel-Art Gallon Bottle, Wide-Mouth, Bel-Art Products, a part of SP
  16126-764
 :  F10638-1010
  • Cap Size:
    110 mm
  • Capacity:
    4 L (1 gal.)
  • Cat. No.:
    16126-764
  • Supplier no.:
    106381010
High-density polyethylene bottle features a foam-lined, white polypropylene cap and offers excellent chemical and impact resistance.
  • HDPE construction
  • Foam-lined polypropylene cap
  • Chemically resistant
Wide mouth facilitates pouring and refilling.