Rack/Filling Station, for Headspace Vials, Acrylic, MicroSolv™
Catalog # 97055-006
Supplier: MicroSolv Technology Corp
Specifications
- Description:Rack, Clear, 160mm x 160mm x 30mm, 25 cavities with a diameter of 24mm
- Cat. No.:97055-006
- Supplier no.:95025-R
Specifications
About this item
The vial racks/filling stations are made of acrylic material so you can see into the vial as you fill it
Also, the silicone base makes them an excellent filling station as they will not slide on the bench.