Order Entry
Puerto Rico
ContactUsLinkComponent
Primary Drinking Water Metals Standard, RICCA Chemical Company
Primary Drinking Water Metals Standard, RICCA Chemical Company
Catalog # RCRWAT7-100
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Primary Drinking Water Metals Standard, RICCA Chemical Company
Catalog # RCRWAT7-100
Supplier:  Ricca Chemical
Supplier Number:  RWAT7000-100B
Small Business Enterprise
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Description:
    Primary Drinking Water Metals Standard
  • Size:
    100 ml
  • Cat. No.:
    RCRWAT7-100
  • Supplier no.:
    RWAT7000-100B

Specifications

About this item

This is a multielement solution that was prepared gravimetrically to contain the elements and concentrations stated below.

Barium (Ba) 99.5-100.5 ppm
Chromium (Cr) 9.95-10.05 ppm
Arsenic (As) 9.95-10.05 ppm
Silver (Ag) 9.95-10.05 ppm
Lead (Pb) 9.95-10.05 ppm
Selenium (Se) 4.97-5.03 ppm
Cadmium (Cd) 4.97-5.03 ppm

The uncertainty associated with the certified values is ± 0.5% relative.

The concentrations are confirmed by ICP or ICP-MS.