Multi-Element Standard for ICP, Trace Metals I for Water, Ricca Chemical Company
Catalog # RCRTRACE1-100
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:LDPE bottle
- Size:100 ml
- Elements and concentration:15 elements
- Cat. No.:RCRTRACE1-100
- Supplier no.:RTRACE10-100N
Specifications
About this item
This is a multielement solution that was prepared volumetrically to contain the elements and concentrations stated below
The uncertainty associated with the certified values is ± 0.5% relative. The concentrations are confirmed by ICP or ICP-MS.