Phosphorus Standard, 50 ppm P, Ricca Chemical Company
Catalog # RCRP33050P500A
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:50 ppm
- Size:500 ml
- Cat. No.:RCRP33050P500A
- Contents:NH₄H₂PO₄ in H₂O
- Supplier no.:RP33050P-500A