Sulfur Standard, 20 ppm S in 2% HNO₃, Ricca Chemical Company
Catalog # RCRP33020S-1A
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:20 µg/ml
- Matrix:2% HNO₃
- Size:1 L
- Cat. No.:RCRP33020S-1A
- Element:Sulfur
- Supplier no.:RP33020S-1A