Thorium ICP Standard, 1 ml = 1 mg Th (1,000 ppm Th)Th(NO3)4 in 3% HNO3, Ricca Chemical Company
Catalog # RCPTH1KN-100
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:1000 µg/ml
- Matrix:3% HNO₃
- Size:100 ml
- Cat. No.:RCPTH1KN-100
- Element:Thorium
- Supplier no.:RPTH1KN0-100N