Order Entry
Puerto Rico
ContactUsLinkComponent
Tellurium ICP Standard, 1 ml = 10 mg Te (10000 ppm Te)Te in 25% HCl, Ricca Chemical Company
Tellurium ICP Standard, 1 ml = 10 mg Te (10000 ppm Te)Te in 25% HCl, Ricca Chemical Company
Catalog # RCPTE10KH-100
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Tellurium ICP Standard, 1 ml = 10 mg Te (10000 ppm Te)Te in 25% HCl, Ricca Chemical Company
Catalog # RCPTE10KH-100
Supplier:  Ricca Chemical
Supplier Number:  RPTE10KH-100N
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    10000 µg/ml
  • Matrix:
    25% HCl
  • Size:
    100 ml
  • Cat. No.:
    RCPTE10KH-100
  • Element:
    Tellurium
  • Supplier no.:
    RPTE10KH-100N

Specifications

About this item