Order Entry
Puerto Rico
ContactUsLinkComponent
 
Silicon ICP Standard, 1 ml = 10 mg Si (10,000 ppm Si)Si in 5% HNO₃ / trace HF, Ricca Chemical Company
Catalog # RCPSI10KN-500
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Silicon ICP Standard, 1 ml = 10 mg Si (10,000 ppm Si)Si in 5% HNO₃ / trace HF, Ricca Chemical Company
Catalog # RCPSI10KN-500
Supplier:  Ricca Chemical
Supplier Number:  RPSI10KN-500N
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    10000 µg/ml
  • Matrix:
    5% HNO₃
  • Size:
    500 ml
  • Cat. No.:
    RCPSI10KN-500
  • Element:
    Silicon
  • Supplier no.:
    RPSI10KN-500N

Specifications

About this item