Antimony ICP Standard, 1 ml = 10 mg Sb (10000 ppm Sb)Sb in 20% HCl, Ricca Chemical Company
RCPSB10KH-500
Small Business Enterprise
:
- Concentration:10000 µg/ml
- Matrix:20% HCl
- Size:500 ml
- Cat. No.:RCPSB10KH-500
- Element:Antimony
- Supplier no.:RPSB10KH-500N