Order Entry
Puerto Rico
ContactUsLinkComponent
Diammonium hydrogen orthophosphate 40% (w/v) in water matrix modifier solution
Irritant
Diammonium hydrogen orthophosphate 40% (w/v) in water matrix modifier solution
Catalog # RCMMAP40W-50
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Irritant
Diammonium hydrogen orthophosphate 40% (w/v) in water matrix modifier solution
Catalog # RCMMAP40W-50
Supplier:  Ricca Chemical
Supplier Number:  RMMAP40W-50N
Small Business Enterprise
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Size:
    50 ml
  • Cat. No.:
    RCMMAP40W-50
  • Supplier no.:
    RMMAP40W-50N

Specifications

Specification Test Results

  • Appearance:
    Clear, colorless, odorless

Specification Test Results

About this item

 :
 :132.06 g/mol
Density:1.2100 g/cm³
 :GHS07
Storage Temperature:Ambient
CAS Number:7783-28-0
Formula:(NH₄)₂HPO₄
MDL Number:MFCD00010891