Order Entry
Puerto Rico
ContactUsLinkComponent
Ammonium nitrate 5% in nitric acid 2% matrix modifier solution
Ammonium nitrate 5% in nitric acid 2% matrix modifier solution
Catalog # RCMMAN5N-50
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Ammonium nitrate 5% in nitric acid 2% matrix modifier solution
Catalog # RCMMAN5N-50
Supplier:  Ricca Chemical
Supplier Number:  RMMAN5N0-50N
Small Business Enterprise
CAS Number:  

Specifications

  • Size:
    50 ml
  • Cat. No.:
    RCMMAN5N-50
  • Supplier no.:
    RMMAN5N0-50N

Specifications

Specification Test Results

  • Appearance:
    Clear, colorless, odorless

Specification Test Results

About this item

 :
 :80.04 g/mol
UN:1942
ADR:5.1,III
CAS Number:6484-52-2
Formula:NH₄NO₃
MDL Number:MFCD00011425