Order Entry
Puerto Rico
ContactUsLinkComponent
Barium hydroxide saturated solution in water USP Test Solution (TS) pH 12.9
Barium hydroxide saturated solution in water USP Test Solution (TS) pH 12.9
Catalog # RC898-32
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Barium hydroxide saturated solution in water USP Test Solution (TS) pH 12.9
Catalog # RC898-32
Supplier:  Ricca Chemical
Supplier Number:  R0898000-1A
Small Business Enterprise
CAS Number:  

Specifications

  • Size:
    1 L
  • Cat. No.:
    RC898-32
  • Supplier no.:
    R0898000-1A

Specifications

Specification Test Results

  • Appearance:
    Slightly hazy, colorless, odorless

Specification Test Results

About this item

 :
 :171.34
UN:3262
ADR:8,III
CAS Number:17194-00-2
Formula:H₂BaO₂
MDL Number:MFCD00003443