Barium diphenylamine-4-sulfonate 0.1% (w/v) in aqueous solution
Catalog # RC8844
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Size:120 ml
- Cat. No.:RC8844
- Supplier no.:R0884000-120A
Specifications
Specification Test Results
- Appearance:Clear, colorless, odorless
- Suitability for Use:Will change from colorless to reddish-violet at end-point
Specification Test Results
About this item
:
:633.89 g/mol
UN:1564
ADR:6.1,III
CAS Number:6211-24-1
:1300-92-1
Formula:C₂₄H₂₀BaN₂O₆S₂
MDL Number:MFCD00007497