Tropaeolin O 0,04% (w/v) in aqueous solution indicator
Supplier: Ricca Chemical
About this item
Synonyms: CI 14270, Tropaeolin O sodium salt, Sulpho orange, Chrysoin, Sulfo orange, 2,4-Dihydroxyazobenzene-4'-sulphonic acid sodium salt, Sodium 2,4-Dihydroxyazobenzene-4'-sulphonate, Acid Orange 6, Resorcinol Yellow
- CAS Number:547-57-9
- Formula:C₁₂H₉N₂O₅SNa
- MDL Number:MFCD00007499
- Molecular Weight:316.26 g/mol
- Boiling Pt:100 °C
- Melting Pt:0.0 °C
- Density:1.0010 g/cm³
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Synonyms: CI 14270, Tropaeolin O sodium salt, Sulpho orange, Chrysoin, Sulfo orange, 2,4-Dihydroxyazobenzene-4'-sulphonic acid sodium salt, Sodium 2,4-Dihydroxyazobenzene-4'-sulphonate, Acid Orange 6, Resorcinol Yellow
- CAS Number:547-57-9
- Formula:C₁₂H₉N₂O₅SNa
- MDL Number:MFCD00007499
- Molecular Weight:316.26 g/mol
- Boiling Pt:100 °C
- Melting Pt:0.0 °C
- Density:1.0010 g/cm³
- Storage Temperature:Ambient
Specifications for Product Family
Specification Test Results
- Appearance:Clear, orange, odorless
- Suitability for Use:Yellow (pH 12.6)
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



