


Sodium hydroxide solution, strong R
Supplier: Ricca Chemical
Certificates
About this item
:
Boiling Pt:~ 100 °C
Melting Pt:~ 0 °C
Density:1.2500 g/cm³
Storage Temperature:Ambient
UN:1824
ADR:8,II
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 723):10.50 ± 0.05 N at 20.0 °C
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_R7481000
SDS
VWR GHS Compliance Letter (US)