


Sodium hydroxide - EDTA solution 10 N
Supplier: Ricca Chemical
Certificates
About this item
:
Boiling Pt:~ 100 °C
Density:1.4000 g/cm³
Storage Temperature:Ambient
UN:1824
ADR:8,II
:GHS05
:GHS07
:GHS08
Specification Test Results
- Ammonia Electrode:No detectable Ammonia
- Appearance:Clear, colorless, alcohol odor
- Assay at 20 °C (traceable to NIST SRM 723):10.0 ± 0.1 N at 20.0 °C