Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tri-Sodium citrate solutions
tri-Sodium citrate 10% (w/w) in aqueous solution
tri-Sodium citrate 10% (w/w) in aqueous solution
RC7230-16
:
Ricca Chemical
Small Business Enterprise
:
tri-Sodium citrate 10% (w/w) in aqueous solution
RC7230-16
:
Ricca Chemical
:
R7230000-500A
Small Business Enterprise
:
Pack type:
Poly bottle
Size:
500 ml
Appearance:
Clear, colorless, odorless
:
~ 100 °C
:
1.0630 g/cm³
:
6132-04-3