Sodium metaarsenite 0.5% (w/v) in aqueous solution
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:7784-46-5
:
Formula:NaAsO₂
MDL Number:MFCD00003472
:129.91
UN:2027
ADR:6.1,II
Specification Test Results
- Appearance:Clear, colorless, odorless