Sodium acetate 13.6% (w/v) in aqueous solution USP Test Solution (TS)
Catalog # RC7113-16
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Poly bottle
- Size:500 ml
- Cat. No.:RC7113-16
- Supplier no.:R7113000-500A
Specifications
Specification Test Results
- Appearance:Clear, colorless, acetate odor
- Assay:13.6 ± 0.5 % (w/v) Sodium Acetate Trihydrate
Specification Test Results
About this item
:
:82.03 g/mol
Boiling Pt:~ 100 °C
Melting Pt:~ 0 °C
Density:1.0400 g/cm³
Storage Temperature:Ambient
CAS Number:127-09-3
Formula:H₃CCOONa
MDL Number:MFCD00012459