



Silver nitrate R1 Reag. Ph. Eur. , 42.5 g/L (0.250 N)
Catalog # RC7040-16
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:500 ml
- Cat. No.:RC7040-16
- Supplier no.:R7040000-500C
Specifications
Specification Test Results
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 999):42.50 ± 0.02 g/L
Specification Test Results
About this item
:
:169.87 g/mol
Boiling Pt:~ 100 °C
Melting Pt:~ 0 °C
Density:1.0400 g/cm³
:GHS08
:GHS05
:GHS09
:GHS06
Storage Temperature:Ambient
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414