Silver nitrate 0.100 N (N/10) in denatured alcohol
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:7761-88-8
:
Formula:AgNO₃
MDL Number:MFCD00003414
:169.87 g/mol
UN:1493
ADR:5.1,II
Specification Test Results
- Appearance:Clear, colorless, alcohol odor
- Assay at 20 °C (traceable to NIST SRM 999):0.1000 ± 0.0001 N at 20.0 °C
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_R7028000
SDS
VWR GHS Compliance Letter (US)