Ammonium metavanadate USP Test Solution (TS)
Catalog # RC698.3-16
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Size:500 ml
- Cat. No.:RC698.3-16
- Supplier no.:R0698300-500A
Specifications
Specification Test Results
- Appearance:Clear, yellow, odorless
Specification Test Results
About this item
:
:116.98 g/mol
UN:2859
ADR:6.1,II
CAS Number:7803-55-6
Formula:NH₄VO₃
MDL Number:MFCD00011430