Order Entry
Puerto Rico
ContactUsLinkComponent
Ammonium Standard, 1 ml = 0.1 mg NH₄+, 100 ppm NH₄+, Ricca Chemical Company
Ammonium Standard, 1 ml = 0.1 mg NH₄+, 100 ppm NH₄+, Ricca Chemical Company
Catalog # RC692.5-100
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Ammonium Standard, 1 ml = 0.1 mg NH₄+, 100 ppm NH₄+, Ricca Chemical Company
Catalog # RC692.5-100
Supplier:  Ricca Chemical
Supplier Number:  R0692500-100A
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    100 ppm
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Cat. No.:
    RC692.5-100
  • Contents:
    NH₄Cl in H₂O
  • Supplier no.:
    R0692500-100A

Specifications

About this item