Silver nitrate 1.7% (w/v) in aqueous solution
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:7761-88-8
:
Formula:AgNO₃
MDL Number:MFCD00003414
:169.87 g/mol
UN:1493
ADR:5.1,II
Specification Test Results
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 999):1.70 ± 0.02 % (w/v)