Potassium permanganate 0.63% (w/v) in aqueous solution
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:7722-64-7
:
Formula:KMnO₄
MDL Number:MFCD00011364
:158.03 g/mol
UN:1490
ADR:5.1,II
Specification Test Results
- Appearance:Clear, purple, odorless