Ammonia Buffer, for Hardness Testing, Ricca Chemical Company
Catalog # RC616-1
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Size:1 gal.
- Cat. No.:RC616-1
- Supplier no.:R0616000-4A
Specifications
About this item
Ammonium chloride/sulfide/hydroxide buffer, pH 10.0 for water hardness testing