Potassium fluoride 40% (w/w) in aqueous solution
Catalog # RC6104-5
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Size:20 L
- Cat. No.:RC6104-5
- Supplier no.:R6104000-20F
Specifications
Specification Test Results
- Appearance:Clear, light pink
- Specific Gravity:1.40 ± 0.01 at 25.0 °C
Specification Test Results
About this item
:
:58.1 g/mol
UN:1812
ADR:6.1,III
CAS Number:7789-23-3
Formula:KF
MDL Number:MFCD00011398