Potassium hexachloroplatinate(IV) 0.2% (w/v) in hydrochloric acid 10%
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:16921-30-5
:
Formula:K₂PtCl₆
MDL Number:MFCD00011389
ADR:6.1,II
:485.99 g/mol
UN:3290
Specification Test Results
- Appearance:Clear, light yellow, odorless to slightly pungent odor