ContactUsLinkComponent
Nitrogen Standard, 1 ml = 0.01 mg N, 10 ppm N as Nitrate (44.27 ppm NO₃⁻), Ricca Chemical Company
Nitrogen Standard, 1 ml = 0.01 mg N, 10 ppm N as Nitrate (44.27 ppm NO₃⁻), Ricca Chemical Company
Supplier:  Ricca Chemical
Certificates
undefined
Nitrogen Standard, 1 ml = 0.01 mg N, 10 ppm N as Nitrate (44.27 ppm NO₃⁻), Ricca Chemical Company
Supplier:  Ricca Chemical