Order Entry
United States
ContactUsLinkComponent
N-(1-Naphthyl)ethylenediamine dihydrochloride USP Test Solution (TS)
N-(1-Naphthyl)ethylenediamine dihydrochloride USP Test Solution (TS)
Catalog # RC5231-16
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
N-(1-Naphthyl)ethylenediamine dihydrochloride USP Test Solution (TS)
Catalog # RC5231-16
Supplier:  Ricca Chemical
Supplier Number:  R5231000-500C
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass bottle
  • Size:
    500 mL
  • Cat. No.:
    RC5231-16
  • Supplier no.:
    R5231000-500C

Specifications

Specification Test Results

  • Appearance:
    Clear, greenish-yellow to slight brown, Acetone odor

Specification Test Results

About this item

 :
 :259.18 g/mol
Boiling Pt:~ 60 °C
Density:0.8526 g/cm³
Flash Pt:–18 °C (Acetone)
Storage Temperature:Refrigerator
UN:1090
ADR:3,II
 :GHS08
 :GHS07
 :GHS02
CAS Number:1465-25-4
Formula:C₁₂H₁₆Cl₂N₂
MDL Number:MFCD00012556