Magnesium Standard, 1 ml = 0.1 mg Mg, 100 ppm Mg, Suitable for Ion Chromatography, Ricca Chemical Company
Catalog # RC4495-100
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:100 µg/ml
- Matrix:H₂O
- Size:100 ml
- Cat. No.:RC4495-100
- Element:Magnesium
- Supplier no.:R4495000-100A