$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Lead(II) acetate
Lead(II) acetate 9.5% (w/v) in aqueous solution USP Test Solution (TS)
Lead(II) acetate 9.5% (w/v) in aqueous solution USP Test Solution (TS)
:
Ricca Chemical
Lead(II) acetate 9.5% (w/v) in aqueous solution USP Test Solution (TS)
:
Ricca Chemical
:
301-04-2
:
:
(H₃CCOO)₂Pb
:
MFCD00012452
:
325.29 g/mol
:
1616
:
6.1,III
Appearance:
Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 915):
9.50 ± 0.04 % (w/v)
ghs_R4293000.pdf
VWR GHS Compliance Letter (US)